| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011822.hg.1 |
ENST00000308420.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011822.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011822.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011822.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011822.hg.1 |
TC09000889.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011825.hg.1 |
ENST00000308420.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011825.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011825.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011825.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011825.hg.1 |
TC09000889.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011823.hg.1 |
ENST00000308420.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011823.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011823.hg.1 |
TC09000889.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011824.hg.1 |
ENST00000308420.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011824.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011824.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011824.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011824.hg.1 |
TC09000889.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164923.hg.1 |
TC0900009456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900164926.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164926.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164926.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164926.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164926.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164926.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164926.hg.1 |
TC0900009456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900164924.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164924.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164924.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164924.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164924.hg.1 |
TC0900009456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900164927.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164927.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164927.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164927.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164927.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164927.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164927.hg.1 |
TC0900009456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009456.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
ENST00000381627.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
ENST00000416837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
NM_005059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
NM_134441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164925.hg.1 |
TC0900009456.hg.1 |
Clariom D TC |