Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000254521.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000367915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000463037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000466176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000466176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000466176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000466176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000466176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000466176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000466176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000466176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000470195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000470195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000470195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000470195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000470195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000470195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000484251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000484251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000484251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000484251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000484251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000485405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000488656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000488656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000488656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000494450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000494450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000494450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000494450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
ENST00000494450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304512.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304512.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304512.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304512.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304512.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304512.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304512.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_001304513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010389.hg.1 |
NM_016371.3 |
Refseq |