| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124526 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103468 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103492 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103506 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103479 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103487 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103463 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103495 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103530 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124603 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103512 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124570 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103483 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124535 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103519 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103521 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103457 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103473 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103465 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103469 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103462 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103502 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103535 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124543 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124525 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124598 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103515 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124554 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124563 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103527 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103503 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103464 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103482 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103510 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103528 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103529 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103522 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103476 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103488 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103481 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103461 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103467 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103486 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103534 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124547 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103516 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103478 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124605 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124562 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103460 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103524 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124569 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124561 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103493 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124577 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103505 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103497 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103475 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
17433 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 8x60K |
22681 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
130604 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
834755 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
AK292291.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
AK292367.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
AY834277.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
BC014450.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
BC029858.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
CR590063.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
CR595324.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
NM_006992.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
NM_201650.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
188614 |
U72508.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
181879 |
AK022184.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
181879 |
BC011853.2 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
181879 |
BC020208.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
128185 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
397950 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P12199 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P12199 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
10728 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P132383 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P132383 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
14424 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_18_P10068454 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P12199 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_23_P12199 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103528 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103527 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124569 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103519 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124588 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124589 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103506 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103522 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124577 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103529 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103510 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103521 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103535 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124597 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103530 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124570 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124603 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103505 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124598 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103524 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124590 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103516 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124605 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103512 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103527 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103524 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124589 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103516 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124570 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103529 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103528 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124588 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103512 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103522 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103530 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124590 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103506 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103510 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103521 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103535 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124569 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124597 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103505 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124603 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124598 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124605 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103519 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124577 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_23_P12199 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P132383 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103527 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103511 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103526 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103531 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103522 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103536 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103516 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103506 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103519 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103504 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103505 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103514 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103521 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103524 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103529 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103535 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103538 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103525 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103530 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103484 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15042024 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15042031 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103520 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15042033 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103523 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15042032 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103522 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103481 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103482 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103521 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103483 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P00034027 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P132383 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P132383 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P12199 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
162776 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
50141 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P17498158 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P132383 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P17497631 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00034027 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103410 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103433 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103421 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103525 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103501 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103458 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103431 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103457 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103462 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103493 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103536 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103529 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103469 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103521 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103506 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103522 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103476 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103532 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103419 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103503 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103484 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103486 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103472 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103454 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103482 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103524 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103429 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103511 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103460 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103408 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103434 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103504 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103480 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103446 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103445 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103473 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103438 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103448 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103538 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103488 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103465 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103516 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103418 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103481 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103426 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103500 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103427 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103505 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103490 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103443 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103412 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103478 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103487 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103441 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103519 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103530 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103424 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103528 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103495 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103498 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103514 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15042028 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103414 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103467 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103450 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103534 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P132383 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00034027 |
- |
- |