Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00370621_m1
          See other CLEC12A GE Assays ›
          Gene Symbol:
          CLEC12A
          Gene Name:
          C-type lectin domain family 12 member A
          Gene Aliases:
          CD371, CLL-1, CLL1, DCAL-2, MICL
          Chromosome Location:
          Chr.12: 9951316 - 9995209 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00370621_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CLEC12A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001207010.1 NP_001193939.1 4 - 5 435 87
          NM_001300730.1 NP_001287659.1 3 - 4 562 87
          NM_138337.5 NP_612210.4 3 - 4 562 87
          NM_201623.3 NP_963917.2 2 - 3 463 87
          XM_005253324.2 XP_005253381.1 4 - 5 449 87
          XM_006719033.3 XP_006719096.1 4 - 5 430 87
          XM_006719035.3 XP_006719098.1 4 - 5 430 87
          XM_006719036.3 XP_006719099.1 4 - 5 430 87
          XM_011520570.1 XP_011518872.1 5 - 6 477 87
          XM_011520571.2 XP_011518873.2 3 - 4 329 87
          XM_011520573.2 XP_011518875.1 4 - 5 430 87
          GenBank mRNA AF247788.1 - 3 - 4 543 87
          AK314001.1 - 3 - 4 562 87
          AY426759.1 - 3 - 4 374 87
          AY498550.1 - 3 - 4 374 87
          AY498551.1 - 2 - 3 275 87
          AY498552.1 - 2 - 3 645 87
          AY547296.1 - 4 - 5 435 87
          BC027967.1 - 2 - 3 737 87
          BC063424.1 - 3 - 4 536 5219159 87
          BC126289.1 - 3 - 4 556 8992005 87
          BC126291.1 - 3 - 4 556 8992007 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200006779.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12003207.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CLEC12A

          Gene Ontology Categories:

          Process(es)

          carbohydrate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000304361.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000350667.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000350667.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000350667.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000350667.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000350667.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000350667.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000355690.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000396507.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000396507.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000396507.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000396507.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000396507.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000434319.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000449959.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000543839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000543839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000543839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000543839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000543839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000543839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000543839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 ENST00000543839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001207010.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_001300730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_138337.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006779.hg.1 NM_201623.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline