| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013190.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013190.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013190.hg.1 |
ENST00000447793.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013190.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013190.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013190.hg.1 |
TC0X000994.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009549.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
ENST00000276062.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
ENST00000377811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
NM_001135998.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
NM_019056.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00165157.hg.1 |
TC0X00009549.hg.1 |
Clariom D TC |