| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013717 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659265 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013695 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659257 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013738 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659319 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013762 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013765 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659315 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659307 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659326 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659305 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659258 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659277 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659267 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659316 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013679 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659272 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013678 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659259 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659286 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013728 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013701 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013683 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659310 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013712 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659249 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013739 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659252 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659281 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013733 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013737 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013680 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659311 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659248 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013763 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013727 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013687 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17166739 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659268 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013730 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659283 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013736 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013748 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659260 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013703 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659255 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013746 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013722 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013760 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659312 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013692 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659278 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013724 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659266 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659251 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013751 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013704 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659301 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
705065 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
337711 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P102694 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P102694 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
29907 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P200561 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P113156 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P113156 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
24305 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
11729 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
50762 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
60484 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
33544 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
24379 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
43997 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
47162 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
17092 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
47030 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P03469310 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_14_P200561 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P03469310 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P102694 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_23_P102694 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659277 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013691 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013710 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013722 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659272 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013687 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013704 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659257 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659251 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659249 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659274 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659253 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013709 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659268 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659259 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659252 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659266 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659265 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013701 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659267 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659278 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659258 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659255 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013717 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013703 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013695 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659272 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013691 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013722 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659252 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659258 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013687 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659253 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659266 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659249 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013695 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659278 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659268 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013704 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659257 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659265 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013709 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659259 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659251 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013710 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659267 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013717 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659255 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013701 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659274 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659277 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013703 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_23_P102694 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P200561 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P113156 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013704 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013708 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013701 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013715 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013694 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013722 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013699 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013691 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013697 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013720 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013717 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013710 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013703 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013706 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013713 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P200561 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P113156 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P113156 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P200561 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P102694 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
109310 |
AK122875.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
109310 |
AL833295.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
109310 |
BC130569.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
109310 |
NM_001007122.2 |
refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
49587 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
62083 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P21047505 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013706 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013689 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013683 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013713 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013745 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013687 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013733 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013678 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013703 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013727 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013752 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013765 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013699 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013679 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P17166739 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013730 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013753 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013720 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013715 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013756 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013761 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013694 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013722 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013735 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013725 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013747 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P17166738 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013701 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013743 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013681 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013750 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013717 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013739 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013691 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013758 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013710 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013697 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P17166741 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013708 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P21047505 |
- |
- |