| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013531 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013499 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659108 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659102 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659135 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659106 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17166730 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013543 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659082 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659126 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659111 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659096 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659125 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013530 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659098 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013522 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659133 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013508 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013552 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659092 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013540 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659080 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013532 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659103 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659122 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013502 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659134 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659136 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659123 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013534 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659131 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659120 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659130 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659097 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013519 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659128 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659117 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659079 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659127 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659112 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659138 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013537 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11013510 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659100 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659119 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659084 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659137 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659110 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31659101 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P385843 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P385843 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
38617 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P106962 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P106962 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
26472 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P41238828 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P34639252 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_14_P106962 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P385843 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_23_P385843 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659117 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659120 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659096 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659108 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013510 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013543 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013507 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659119 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659099 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013530 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659122 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659110 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013511 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659091 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659111 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659101 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013532 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659106 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659100 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659087 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013502 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013519 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659102 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659094 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013534 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013537 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659092 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659106 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013511 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659100 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013519 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659117 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013534 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659091 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659122 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013530 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659111 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659101 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659110 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013532 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659096 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659092 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659087 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013537 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013543 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659099 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659108 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659094 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659119 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659120 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013502 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013507 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31659102 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11013510 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_23_P385843 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P106962 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013523 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013532 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013529 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013528 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013509 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013525 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013536 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013512 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013518 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013539 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013507 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013515 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013514 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013542 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11013520 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P106962 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P106962 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P385843 |
- |
- |
| Custom Human Agilent Array 11k, Chromosome 6, Expression |
10115 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
AK126566.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
AK225159.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
AK291860.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
BC011792.2 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
BC014786.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
BC017408.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
BC098453.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR591368.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR593921.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR598218.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR600133.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR600240.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR603672.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR610523.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR611035.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR615338.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR620644.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR622906.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
CR625294.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
NM_001398.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120707 |
U16660.1 |
genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
6349 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P21047413 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013505 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013520 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013514 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013528 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013499 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013553 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013548 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013497 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013509 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013545 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013502 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013532 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013541 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013555 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013525 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013551 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013523 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013557 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013539 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013534 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013542 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013536 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013558 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013515 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013518 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013511 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013504 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11013507 |
- |
- |