| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040981.hg.1 |
ENST00000271139.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040981.hg.1 |
ENST00000319928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040981.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040981.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040981.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040981.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040981.hg.1 |
TC01006354.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040985.hg.1 |
ENST00000271139.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040985.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040985.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040985.hg.1 |
TC01006354.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100241999.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241999.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241999.hg.1 |
TC0100018438.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100241995.hg.1 |
TC0100018438.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100242001.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242001.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242001.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242001.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242001.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242001.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242001.hg.1 |
TC0100018438.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000319928.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
ENST00000477318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018438.hg.1 |
NM_201403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242000.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242000.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242000.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242000.hg.1 |
ENST00000271139.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242000.hg.1 |
ENST00000371940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242000.hg.1 |
NM_145279.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242000.hg.1 |
TC0100018438.hg.1 |
Clariom D TC |