| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004343.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004343.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004343.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004343.hg.1 |
TC02000285.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200151738.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200151738.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200151738.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200151738.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200151738.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200151738.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200151738.hg.1 |
TC0200007473.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000238892.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
ENST00000486447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007473.hg.1 |
NM_014171.5 |
Refseq |