| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008038.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008038.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008038.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008038.hg.1 |
TC17000646.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008035.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008035.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008035.hg.1 |
TC17000646.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008037.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008037.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008037.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17008037.hg.1 |
TC17000646.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008211.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160726.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160726.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160726.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160726.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160726.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160726.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160726.hg.1 |
TC1700008211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700160725.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160725.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160725.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160725.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160725.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160725.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160725.hg.1 |
TC1700008211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
ENST00000570453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160727.hg.1 |
TC1700008211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700160722.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160722.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160722.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160722.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160722.hg.1 |
TC1700008211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700160724.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160724.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160724.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160724.hg.1 |
ENST00000328741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160724.hg.1 |
ENST00000513748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700160724.hg.1 |
NM_007225.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700160724.hg.1 |
TC1700008211.hg.1 |
Clariom D TC |