Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00379469_m1
          See other SERPINB9P1 GE Assays ›
          Gene Symbol:
          SERPINB9P1
          Gene Name:
          serpin family B member 9 pseudogene 1
          Gene Aliases:
          SERPINB9P
          Chromosome Location:
          Chr.6: 2854657 - 2876510 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00379469_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SERPINB9P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033851.1 1 - 2 224 131
          GenBank mRNA BC025340.1 - 1 - 2 224 131
          OA988518.1 - 2 - 3 262 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014218.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001219.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC025340.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514367 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514357 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700913 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514373 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514364 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700916 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514451 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15778335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514432 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514358 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514355 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514441 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24700853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04514366 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 124491 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 105975 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 7642 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 732329 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 522758 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 469702 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 54216 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 868049 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 421299 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 256168 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 638606 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 175672 AF228708.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 175672 NM_033257.2 refseq_mrna_human_NM
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P183918 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11575 - -
          Agilent Human Genome CGH Microarray 44B A_14_P102267 - -
          Agilent Human Genome CGH Microarray 44A A_14_P102267 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55720 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P183918 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P183918 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P102267 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P17427824 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17427824 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P102267 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17427815 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 12215 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 96280 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 2887 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 21530 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P24732990 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P31671112 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15525768 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15523470 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15522507 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15520531 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17427824 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P24731641 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17427831 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P24732872 - -
          Agilent Human 1B Microarray G4111A A_32_P183918 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15522507 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P24731641 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15523470 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17427824 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15520531 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8123607 - -
          Affymetrix Human Gene 1.0 ST Array 8123608 - -
          Affymetrix Human Gene 1.1 ST Array 8123606 - -
          Affymetrix HT HG-U133+ PM Array Plate 239186_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06014499.hg.1 ENST00000420981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014499.hg.1 ENST00000545177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014499.hg.1 NR_033851.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014499.hg.1 TC06001219.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06014501.hg.1 ENST00000420981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014501.hg.1 ENST00000545177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014501.hg.1 NR_033851.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014501.hg.1 TC06001219.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 ENST00000583158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 NR_033851.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 NR_033851.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 NR_033851.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 NR_033851.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014218.hg.1 NR_039788.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 ENST00000583158.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014218.hg.1 NR_039788.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200199.hg.1 TC0600014218.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600200213.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200213.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200213.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200213.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200213.hg.1 TC0600014218.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600200198.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200198.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200198.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200198.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200198.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200198.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200198.hg.1 TC0600014218.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 ENST00000420981.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 ENST00000545177.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 NR_033851.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600200211.hg.1 TC0600014218.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 239186_PM_at - -
          Human Genome U133 Plus 2.0 Array 239186_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239186_at3 - -
          Human Genome U95 Set 56577_PM_at - -
          Human Genome U95 Set 56577_at - -
          Human Genome U133 Set 239186_at - -
          Human Genome U133 Set 239186_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A055212 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A055212 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 186314 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1669122 NR_033851.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1669122 BC025340.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1669122 NR_033851.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1669122 BC025340.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1669122 NR_033851.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1669122 BC025340.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1669122 NR_033851.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1669122 BC025340.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1669122 NR_033851.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1669122 BC025340.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline