Search
Search

RNF113B| Gene Symbol: RNF113B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178861.4 | NP_849192.1 | 1 - 2 | 986 | 77 | |
| GenBank mRNA | AI935007.1 | - | 1 - 2 | 77 | ||
| BC017585.2 | - | 1 - 2 | 986 | 4828542 | 77 | |
| BC025388.1 | - | 1 - 2 | 979 | 4824788 | 77 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09068706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09068708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09068705 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09068704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09068727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09068719 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16662313 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517770 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29517789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16662312 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P297480 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P297480 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 21964 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3740 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P297480 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P297480 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16662305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068718 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517790 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16662313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068704 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068704 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068718 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16662313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517790 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29517787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09068708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16662305 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P297480 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068712 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068719 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068718 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068708 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068704 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068710 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068714 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068725 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068702 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068707 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068713 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068721 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068711 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068705 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09068716 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09068706 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09068704 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16662310 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09068705 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09068707 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16662304 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09068703 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16662307 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16662311 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16662309 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16662308 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P297480 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P491430 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068716 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068702 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16662313 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068719 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068707 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068705 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068704 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068708 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068712 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068717 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068718 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16662304 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068710 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09068714 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_178861_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7972441 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7972440 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7972439 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7972438 | - | - |
| Affymetrix Human Genome U219 Array | 11733483_at | - | - |
| Affymetrix Human Genome U219 Array | 11733483_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009126.hg.1 | ENST00000267291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009126.hg.1 | NM_178861.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009126.hg.1 | TC13000817.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009560.hg.1 | ENST00000267291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009560.hg.1 | ENST00000267291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009560.hg.1 | NM_178861.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009560.hg.1 | NM_178861.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009560.hg.1 | ENST00000267291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009560.hg.1 | ENST00000267291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009560.hg.1 | NM_178861.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009560.hg.1 | NM_178861.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163960.hg.1 | ENST00000267291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163960.hg.1 | NM_178861.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163960.hg.1 | ENST00000267291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163960.hg.1 | NM_178861.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163960.hg.1 | TC1300009560.hg.1 | Clariom D TC |
| Human Genome U95 Set | 85013_PM_at | - | - |
| Human Genome U95 Set | 85013_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049763 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 128044 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_37059780-S | NM_178861.4 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_37059780-S | AF539427.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_37059780-S | AI935007.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_37059780-S | BC017585.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_37059780-S | BC025388.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_20956-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006420170 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006420170 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1500333 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_20956 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1768795 | BC025388.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768795 | NM_178861.4 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768795 | AF539427.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768795 | AI935007.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768795 | BC017585.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768795 | BC025388.1 | human_transcriptome |