| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022280.hg.1 |
TC05001638.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022280.hg.1 |
ENST00000231461.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022280.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022280.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022280.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022282.hg.1 |
TC05001638.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022282.hg.1 |
ENST00000231461.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022282.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022282.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022282.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022284.hg.1 |
TC05001638.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022284.hg.1 |
ENST00000231461.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022284.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022284.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000507360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000507360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000507360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000507360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011579.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180163.hg.1 |
TC0500011579.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500180166.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180166.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180166.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180166.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180166.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180166.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180166.hg.1 |
TC0500011579.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180164.hg.1 |
TC0500011579.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
ENST00000231461.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
ENST00000451528.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
ENST00000523381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
NM_005668.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
NM_175052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180161.hg.1 |
TC0500011579.hg.1 |
Clariom D TC |