| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006115.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006115.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006115.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006115.hg.1 |
TC11000513.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006116.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006116.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006116.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006116.hg.1 |
TC11000513.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000532591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000532591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000532591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
ENST00000532591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154902.hg.1 |
TC1100007775.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000527161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000532591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000532591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000532591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
ENST00000532591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007775.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
ENST00000320202.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
ENST00000412599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
NR_026946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154903.hg.1 |
TC1100007775.hg.1 |
Clariom D TC |