| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018932.hg.1 |
TC03001094.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018932.hg.1 |
ENST00000296327.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018932.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018929.hg.1 |
TC03001094.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018929.hg.1 |
ENST00000296327.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03018929.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000471430.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000471430.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000471430.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000471430.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000476129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000476129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000476129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000476129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000416660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000428985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000442203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000471430.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000471430.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000471430.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000471430.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000472653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000476129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000476129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000476129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000476129.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013918.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000296327.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000475271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000475672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000484407.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
ENST00000492794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
NM_152672.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206191.hg.1 |
TC0300013918.hg.1 |
Clariom D TC |