Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00381885_m1
          See other MGC40069 GE Assays ›
          Gene Symbol:
          MGC40069
          Gene Name:
          uncharacterized protein MGC40069
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00381885_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MGC40069
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC032242.1 - 2 - 3 282 115

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09160589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09160595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16684434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09160593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16684435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16684440 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16684382 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09160590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16684439 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29617531 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3715843 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 1126 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3715843 - -
          Agilent Human 1B Microarray G4111A A_32_P706303 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7973302 - -
          Affymetrix Human Gene 1.0 ST Array 7973299 - -
          Affymetrix Human Gene 1.0 ST Array 7973301 - -
          Affymetrix Human Gene 1.0 ST Array 7973300 - -
          Affymetrix Human Gene 1.1 ST Array 7973298 - -
          Affymetrix HT HG-U133+ PM Array Plate 243602_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1555073_PM_at - -
          Human Genome U133 Plus 2.0 Array 215797_at - -
          Human Genome U133 Plus 2.0 Array 1555073_at - -
          Human Genome U133 Plus 2.0 Array 215797_PM_at - -
          Human Genome U133 Plus 2.0 Array 243602_at - -
          Human Genome U133 Plus 2.0 Array 243602_PM_at - -
          Human Genome U133 Plus 2.0 Array 1555073_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243602_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555073_at5 - -
          Human Genome U95 Set 83750_at - -
          Human Genome U95 Set 83750_PM_at - -
          Human Genome U133 Set 243602_at - -
          Human Genome U133 Set 243602_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 184553 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1664248 BC032242.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1754204 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1664248 BC032242.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1754204 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1754204 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1664248 BC032242.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1664248 BC032242.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1664248 BC032242.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1664248 BC032242.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline