Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00382267_m1
          See other LINC01312 GE Assays ›
          Gene Symbol:
          LINC01312
          Gene Name:
          long intergenic non-protein coding RNA 1312
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 133821147 - 133853992 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00382267_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01312
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027030.1 2 - 3 723 111
          GenBank mRNA BC032958.2 - 2 - 3 723 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600009504.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001000.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291067 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291076 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291082 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291078 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291072 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291074 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090896 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291085 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291106 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290907 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291069 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090913 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290913 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090921 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291089 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05090725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25290939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25291056 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P333057 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P333057 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P333057 - -
          Agilent Human 1B Microarray G4111A A_32_P462941 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1552900_PM_a_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1552899_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06011811.hg.1 TC06001000.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06011811.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011811.hg.1 NR_027030.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011813.hg.1 TC06001000.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06011813.hg.1 NR_027030.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011812.hg.1 TC06001000.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06011812.hg.1 NR_027030.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165226.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165226.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165226.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165226.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165226.hg.1 TC0600009504.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600165227.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165227.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165227.hg.1 TC0600009504.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009504.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 ENST00000412745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 ENST00000456347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 NR_027030.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600165225.hg.1 TC0600009504.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1552900_a_at - -
          Human Genome U133 Plus 2.0 Array 1552899_at - -
          Human Genome U133 Plus 2.0 Array 1552899_PM_at - -
          Human Genome U133 Plus 2.0 Array 1552900_PM_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552899_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552900_a_at3 - -
          Human Genome U95 Set 52293_at - -
          Human Genome U95 Set 52293_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032094 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032095 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032094 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032095 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 226280 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline