Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00382437_g1
          See other TMEM277P GE Assays ›
          Gene Symbol:
          TMEM277P
          Gene Name:
          transmembrane protein 277, pseudogene
          Gene Aliases:
          LINC01869
          Chromosome Location:
          Chr.19: 50804901 - 50818878 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00382437_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMEM277P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038359.1 2 - 3 194 56
          NR_197389.1 5 - 6 669 56
          NR_197390.1 4 - 5 584 56
          GenBank mRNA BC033237.1 - 2 - 3 194 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900008604.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000769.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC033237.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P405282 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 30381 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 35747 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P405282 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P34593107 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8030750 - -
          Affymetrix HT HG-U133+ PM Array Plate 1554222_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR19010904.hg.1 ENST00000326989.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19010904.hg.1 NR_038359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19010904.hg.1 TC19000769.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19010902.hg.1 ENST00000326989.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19010902.hg.1 NR_038359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19010902.hg.1 TC19000769.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19010903.hg.1 ENST00000326989.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19010903.hg.1 NR_038359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19010903.hg.1 TC19000769.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 ENST00000593632.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 ENST00000593632.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167194.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167194.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167194.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167194.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167194.hg.1 TC1900008604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900167191.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167191.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167191.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167191.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167191.hg.1 TC1900008604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900167193.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167193.hg.1 ENST00000593632.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167193.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167193.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167193.hg.1 ENST00000593632.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167193.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167193.hg.1 TC1900008604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 ENST00000593632.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 ENST00000593632.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008604.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167186.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167186.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167186.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167186.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167186.hg.1 TC1900008604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900167188.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167188.hg.1 ENST00000593632.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167188.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167188.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167188.hg.1 ENST00000593632.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167188.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167188.hg.1 TC1900008604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900167196.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167196.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167196.hg.1 ENST00000326989.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900167196.hg.1 NR_038359.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900167196.hg.1 TC1900008604.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1554222_at - -
          Human Genome U133 Plus 2.0 Array 1554222_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554222_at9 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A012930 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A012930 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 227412 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1798964 NR_038359.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1798964 BC033237.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1787731 NR_038359.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1787731 BC033237.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1787731 NR_038359.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1787731 BC033237.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1798964 NR_038359.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1798964 BC033237.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1787731 NR_038359.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1787731 BC033237.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1798964 NR_038359.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1798964 BC033237.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1798964 NR_038359.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1798964 BC033237.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1787731 NR_038359.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1787731 BC033237.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline