| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000535353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000535353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000535353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000370292.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000439173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000599845.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008754.hg.1 |
ENST00000617158.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000356911.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000535353.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000535353.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000535353.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NM_001129828.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011132.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175438.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175438.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175438.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175438.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175438.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175438.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175438.hg.1 |
TC0X00011132.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00175439.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175439.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175439.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175439.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175439.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175439.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175439.hg.1 |
TC0X00011132.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00175440.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175440.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175440.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175440.hg.1 |
ENST00000415810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175440.hg.1 |
ENST00000598036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175440.hg.1 |
NR_130638.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175440.hg.1 |
TC0X00011132.hg.1 |
Clariom D TC |