Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00384077_m1
          See other MYDGF GE Assays ›
          Gene Symbol:
          MYDGF
          Gene Name:
          myeloid derived growth factor
          Gene Aliases:
          C19orf10, EUROIMAGE1875335, IL25, IL27, IL27w, R33729_1, SF20
          Chromosome Location:
          Chr.19: 4657545 - 4670403 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00384077_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MYDGF
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019107.3 NP_061980.1 2 - 3 292 82
          XM_017026987.1 XP_016882476.1 2 - 3 280 82
          GenBank mRNA AF282264.1 - 2 - 3 223 82
          AK098829.1 - 2 - 3 231 82
          AL365373.1 - 2 - 3 214 82
          AL365374.1 - 2 - 3 214 82
          BC003639.2 - 2 - 3 210 82
          BC010129.2 - 2 - 3 259 3953027 82
          BC014655.1 - 2 - 3 230 4562455 82
          BG484758.1 - 2 - 3 293 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900009357.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001067.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MYDGF

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          apoptotic process
          positive regulation of phosphatidylinositol 3-kinase signaling
          IRE1-mediated unfolded protein response
          negative regulation of apoptotic process
          positive regulation of MAPK cascade
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of protein kinase B signaling
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19014600.hg.1 ENST00000262947.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19014600.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19014600.hg.1 TC19001067.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19014601.hg.1 ENST00000262947.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19014601.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19014601.hg.1 TC19001067.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19014602.hg.1 ENST00000262947.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19014602.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19014602.hg.1 TC19001067.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19014603.hg.1 ENST00000262947.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19014603.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19014603.hg.1 TC19001067.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19014604.hg.1 ENST00000262947.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19014604.hg.1 NM_019107.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19014604.hg.1 TC19001067.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 TC1900009357.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174775.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900174778.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174778.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174778.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900174778.hg.1 TC1900009357.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900174778.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174778.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174778.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 TC1900009357.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 ENST00000262947.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 ENST00000596031.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 ENST00000599630.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900174776.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900174782.hg.1 NM_019107.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900174782.hg.1 TC1900009357.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900174782.hg.1 NM_019107.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 147923 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline