| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015755.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015755.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015755.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015755.hg.1 |
TC09001226.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015754.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015754.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015754.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015754.hg.1 |
TC09001226.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171711.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171711.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171711.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171711.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171711.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171711.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171711.hg.1 |
TC0900010482.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171709.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171709.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171709.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171709.hg.1 |
ENST00000341700.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171709.hg.1 |
ENST00000464095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171709.hg.1 |
NM_004297.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171709.hg.1 |
TC0900010482.hg.1 |
Clariom D TC |