Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00393094_m1
          See other ZNRF3 GE Assays ›
          Gene Symbol:
          ZNRF3
          Gene Name:
          zinc and ring finger 3
          Gene Aliases:
          BK747E2.3, RNF203
          Chromosome Location:
          Chr.22: 28883572 - 29057488 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00393094_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNRF3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001206998.2 NP_001193927.1 8 - 9 2963 57
          NM_032173.4 NP_115549.2 8 - 9 2561 57
          GenBank mRNA AB051436.1 - 8 - 9 2633 57
          AK025710.1 - 3 - 4 1955 57
          AK096397.1 - 8 - 9 2561 57
          BC009252.2 - 3 - 4 1892 57
          BC021570.1 - 3 - 4 1920 57
          BC069019.1 - 3 - 4 1891 57
          BC094857.1 - 3 - 4 1891 57
          CR456397.1 - 8 - 9 2541 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200009246.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000189.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          Wnt signaling pathway
          protein ubiquitination
          negative regulation of Wnt signaling pathway
          Wnt receptor catabolic process
          limb development
          regulation of canonical Wnt signaling pathway
          stem cell proliferation
          negative regulation of canonical Wnt signaling pathway
          regulation of non-canonical Wnt signaling pathway
          negative regulation of non-canonical Wnt signaling pathway
          regulation of Wnt signaling pathway, planar cell polarity pathway
          ubiquitin-protein transferase activity
          frizzled binding
          protein binding
          zinc ion binding
          transferase activity
          metal ion binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22003480.hg.1 ENST00000332811.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003480.hg.1 ENST00000402174.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003480.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003480.hg.1 ENST00000462485.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003480.hg.1 ENST00000544604.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003480.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003480.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003480.hg.1 TC22000189.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay TC2200009246.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 ENST00000402174.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 ENST00000406323.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 ENST00000544604.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 NM_001206998.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 NM_032173.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200168379.hg.1 TC2200009246.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline