| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486068 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486084 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486070 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486083 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486059 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486064 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623758 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623773 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486035 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486045 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486073 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486029 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623760 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486057 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623799 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623752 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623781 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623780 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623792 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486047 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623808 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486039 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623804 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623801 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486044 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486071 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623786 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486031 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486052 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486060 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623803 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623806 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623756 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486046 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486066 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486015 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623748 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486069 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486034 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486043 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623802 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486016 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486033 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486061 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623800 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486065 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623783 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623805 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486038 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22623759 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486019 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486013 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02486032 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
46096 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
116939 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P92196 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P92196 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
10782 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P135758 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P135758 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
23031 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
6319 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P58527989 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P00781189 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P00781187 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P92196 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_23_P92196 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623780 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623782 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486065 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486072 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486061 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623786 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623802 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486052 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486050 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623759 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486046 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486066 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486044 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486070 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623781 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486064 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486035 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486059 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486057 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486034 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623760 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623792 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623800 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486032 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486039 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486068 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486069 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486033 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486047 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486042 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623803 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486031 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623783 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623780 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486031 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623781 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486035 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486070 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486068 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623800 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486066 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486069 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486047 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486034 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623786 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623760 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486072 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486061 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486052 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623782 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486057 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623759 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623802 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623792 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486064 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486065 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486044 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486033 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486046 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623783 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486042 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486039 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486059 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486050 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02486032 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22623803 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_23_P92196 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P135758 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486032 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486039 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486049 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486028 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486046 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486072 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486048 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486075 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486068 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486031 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486042 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486054 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486065 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486061 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486035 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486044 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486057 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486063 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02486051 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P135758 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P135758 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P92196 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
128609 |
AK129864.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
128609 |
NM_000037.3 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
128609 |
NM_020475.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
128609 |
NM_020476.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
128609 |
NM_020477.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
128609 |
NM_020478.3 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
128609 |
NM_020480.3 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
128609 |
X16609.1 |
genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
18074 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00781189 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486014 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486081 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486026 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486028 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486061 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486048 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486022 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486058 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486044 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486018 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486083 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486063 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486039 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486051 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486070 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486035 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486054 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486077 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486024 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486042 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486056 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486016 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486031 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486020 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486068 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486080 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486013 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486072 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486046 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486075 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02486065 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00781187 |
- |
- |