| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000461104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000461104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000461104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007952.hg.1 |
ENST00000373295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007952.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09007952.hg.1 |
TC09000669.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000461104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000461104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000461104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000464092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008816.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
ENST00000373295.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
ENST00000496009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
ENST00000614677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
NM_001012502.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900158977.hg.1 |
TC0900008816.hg.1 |
Clariom D TC |