Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00405789_m1
          See other OBSCN GE Assays ›
          Gene Symbol:
          OBSCN
          Gene Name:
          obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
          Gene Aliases:
          ARHGEF30, UNC89
          Chromosome Location:
          Chr.1: 228208013 - 228381431 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00405789_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OBSCN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001098623.2 NP_001092093.2 56 - 57 15728 96
          NM_001271223.2 NP_001258152.2 67 - 68 18599 96
          NM_052843.3 NP_443075.3 56 - 57 15728 96
          XM_005273287.4 XP_005273344.1 68 - 69 18979 96
          XM_005273291.4 XP_005273348.1 68 - 69 18943 96
          XM_005273298.4 XP_005273355.1 67 - 68 18670 96
          XM_005273307.4 XP_005273364.1 63 - 64 17857 96
          XM_006711822.3 XP_006711885.1 66 - 67 18403 96
          XM_006711823.3 XP_006711886.1 66 - 67 18403 96
          XM_006711827.3 XP_006711890.1 60 - 61 16818 96
          XM_006711829.3 XP_006711892.1 59 - 60 16542 96
          XM_011544297.2 XP_011542599.1 60 - 61 16819 96
          XM_011544299.2 XP_011542601.1 59 - 60 16543 96
          XM_017002443.1 XP_016857932.1 68 - 69 19024 96
          XM_017002444.1 XP_016857933.1 68 - 69 18987 96
          XM_017002445.1 XP_016857934.1 68 - 69 19024 96
          XM_017002446.1 XP_016857935.1 68 - 69 19024 96
          XM_017002447.1 XP_016857936.1 68 - 69 19021 96
          XM_017002448.1 XP_016857937.1 68 - 69 18874 96
          XM_017002449.1 XP_016857938.1 67 - 68 18761 96
          XM_017002450.1 XP_016857939.1 67 - 68 18761 96
          XM_017002451.1 XP_016857940.1 67 - 68 18761 96
          XM_017002452.1 XP_016857941.1 67 - 68 18761 96
          XM_017002453.1 XP_016857942.1 67 - 68 18748 96
          XM_017002454.1 XP_016857943.1 67 - 68 18748 96
          XM_017002455.1 XP_016857944.1 67 - 68 18748 96
          XM_017002456.1 XP_016857945.1 67 - 68 18748 96
          XM_017002457.1 XP_016857946.1 67 - 68 18748 96
          XM_017002458.1 XP_016857947.1 66 - 67 18497 96
          XM_017002459.1 XP_016857948.1 66 - 67 18497 96
          XM_017002460.1 XP_016857949.1 66 - 67 18497 96
          XM_017002461.1 XP_016857950.1 66 - 67 18484 96
          XM_017002462.1 XP_016857951.1 66 - 67 18472 96
          XM_017002463.1 XP_016857952.1 66 - 67 18451 96
          XM_017002464.1 XP_016857953.1 66 - 67 18400 96
          XM_017002465.1 XP_016857954.1 65 - 66 18233 96
          XM_017002466.1 XP_016857955.1 65 - 66 18233 96
          XM_017002467.1 XP_016857956.1 65 - 66 18233 96
          XM_017002468.1 XP_016857957.1 64 - 65 17969 96
          XM_017002469.1 XP_016857958.1 62 - 63 17441 96
          XM_017002470.1 XP_016857959.1 57 - 58 16122 96
          XM_017002471.1 XP_016857960.1 68 - 69 19023 96
          XM_017002472.1 XP_016857961.1 68 - 69 19023 96
          XM_017002473.1 XP_016857962.1 68 - 69 19023 96
          GenBank mRNA AJ002535.1 - 57 - 58 15725 96
          AL832357.1 - 1 - 2 144 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100011861.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OBSCN

          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          multicellular organism development
          regulation of Rho protein signal transduction
          protein localization to M-band
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          sarcomere organization
          regulation of small GTPase mediated signal transduction
          mitophagy in response to mitochondrial depolarization
          protein serine/threonine kinase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          structural constituent of muscle
          ankyrin binding
          titin binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 ENST00000284548.15 Ensembl
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 ENST00000366707.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 ENST00000422127.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 ENST00000570156.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 NM_001098623.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 NM_001271223.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 NM_052843.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 ENST00000284548.15 Ensembl
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 ENST00000366707.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 ENST00000422127.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 ENST00000570156.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 NM_001098623.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 NM_001271223.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 NM_052843.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100187611.hg.1 TC0100011861.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 162718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline