Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00415150_g1
          See other MGC39584 GE Assays ›
          Gene Symbol:
          MGC39584
          Gene Name:
          uncharacterized LOC441058
          Gene Aliases:
          -
          Chromosome Location:
          Chr.21: 9781848 - 9821061 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00415150_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MGC39584
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038377.1 3 - 4 601 106
          GenBank mRNA BC029568.1 - 3 - 4 601 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007547.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC4_gl000193_random000001.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P111996 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P30834 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P111996 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17631 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 34383 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 17314 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P111996 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P30834 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03808908 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03808908 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21696407 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 43481 - -
          Agilent Human 1B Microarray G4111A A_32_P111996 - -
          Agilent Human 1B Microarray G4111A A_32_P30834 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 242822_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 240814_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR4_gl000193_random000002.hg.1 TC4_gl000193_random000001.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_gl000193_random000002.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR4_gl000193_random000003.hg.1 TC4_gl000193_random000001.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_gl000193_random000003.hg.1 NR_038377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR4_gl000193_random000001.hg.1 TC4_gl000193_random000001.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR4_gl000193_random000001.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007547.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151300.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151300.hg.1 TC2100007547.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100151300.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 TC2100007547.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 ENST00000623919.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 ENST00000623933.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151308.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151303.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151303.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151303.hg.1 TC2100007547.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100151303.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151303.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151304.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151304.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151304.hg.1 TC2100007547.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100151304.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151304.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151302.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151302.hg.1 NR_038377.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100151302.hg.1 TC2100007547.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100151302.hg.1 ENST00000623554.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100151302.hg.1 NR_038377.1 Refseq
          Human Genome U133 Plus 2.0 Array 242822_at - -
          Human Genome U133 Plus 2.0 Array 240814_at - -
          Human Genome U133 Plus 2.0 Array 240814_PM_at - -
          Human Genome U133 Plus 2.0 Array 242822_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240814_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 242822_at3 - -
          Human Genome U95 Set 53455_PM_at - -
          Human Genome U95 Set 53455_at - -
          Human Genome U95 Set 75542_at - -
          Human Genome U95 Set 75542_PM_at - -
          Human Genome U95 Set 69054_PM_r_at - -
          Human Genome U95 Set 69054_r_at - -
          Human Genome U95 Set 82915_at - -
          Human Genome U95 Set 82915_PM_at - -
          Human Genome U133 Set 242822_at - -
          Human Genome U133 Set 240814_at - -
          Human Genome U133 Set 240814_PM_at - -
          Human Genome U133 Set 242822_PM_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1747568 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1747568 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1747568 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1747568 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline