| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
232836_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000558175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
ENST00000558175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008401.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
ENST00000558175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
ENST00000558175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159138.hg.1 |
TC2000008401.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000558175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
ENST00000558175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008401.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159136.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159136.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159136.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159136.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159136.hg.1 |
ENST00000557899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159136.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159136.hg.1 |
TC2000008401.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000159135.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159135.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159135.hg.1 |
ENST00000427835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159135.hg.1 |
NR_110635.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159135.hg.1 |
TC2000008401.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
232836_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
232836_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232836_at6 |
- |
- |
| Human Genome U95 Set |
58626_at |
- |
- |
| Human Genome U95 Set |
58626_PM_at |
- |
- |
| Human Genome U133 Set |
232836_at |
- |
- |
| Human Genome U133 Set |
232836_PM_at |
- |
- |