Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00415854_m1
          See other SLC16A9 GE Assays ›
          Gene Symbol:
          SLC16A9
          Gene Name:
          solute carrier family 16 member 9
          Gene Aliases:
          C10orf36, MCT9
          Chromosome Location:
          Chr.10: 59650764 - 59710079 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00415854_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC16A9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001323977.1 NP_001310906.1 2 - 3 168 68
          NM_001323978.1 NP_001310907.1 3 - 4 596 68
          NM_001323979.1 NP_001310908.1 2 - 3 283 68
          NM_001323980.1 NP_001310909.1 2 - 3 513 68
          NM_001323981.1 NP_001310910.1 3 - 4 726 68
          NM_194298.2 NP_919274.1 2 - 3 643 68
          XM_017015883.1 XP_016871372.1 2 - 3 643 68
          XM_017015884.1 XP_016871373.1 3 - 4 976 68
          GenBank mRNA AK125791.1 - 2 - 3 303 68
          BC126238.1 - 1 - 2 232 8991954 68
          BC130567.1 - 1 - 2 232 40146598 68
          DA759951.1 - 2 - 3 168 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010722.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001315.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001323977.1,NM_001323978.1,NM_001323979.1,NM_001323980.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC16A9

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          monocarboxylic acid transport
          urate metabolic process
          transmembrane transport
          protein binding
          monocarboxylic acid transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10016493.hg.1 ENST00000395348.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016493.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016493.hg.1 TC10001315.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10016489.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016489.hg.1 ENST00000395348.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016489.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016489.hg.1 TC10001315.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10016487.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016487.hg.1 ENST00000395348.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016487.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016487.hg.1 TC10001315.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10016492.hg.1 ENST00000395348.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016492.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016492.hg.1 TC10001315.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173060.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173060.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173060.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173060.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173060.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173060.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173060.hg.1 TC1000010722.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173062.hg.1 TC1000010722.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173063.hg.1 TC1000010722.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000173067.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173067.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173067.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173067.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173067.hg.1 TC1000010722.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395347.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010722.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173066.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173066.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173066.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173066.hg.1 ENST00000395348.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173066.hg.1 ENST00000490066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173066.hg.1 NM_194298.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000173066.hg.1 TC1000010722.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline