Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00416124_m1
          See other NSMCE1-DT GE Assays ›
          Gene Symbol:
          NSMCE1-DT
          Gene Name:
          NSMCE1 divergent transcript
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 27268205 - 27290468 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00416124_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NSMCE1-DT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037184.1 3 - 4 227 84
          GenBank mRNA BC044845.1 - 3 - 4 227 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600007310.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000291.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976636 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976640 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976642 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16871388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16871385 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501120 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16871387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09976615 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30501125 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 132583 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 562693 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 877815 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 6923 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 189275 AB007963.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 189275 NM_014774.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 123531 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 725227 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 70710 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3484806 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P882871 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P87191 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 18834 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 18965 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 31908 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P20436995 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P20436995 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3484806 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P882871 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P87191 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P20436995 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P882871 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P87191 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 87651 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20436995 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03132195 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03132216 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40595412 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20436992 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03132212 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20436997 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03132210 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40595424 - -
          Agilent Human 1B Microarray G4111A A_32_P87192 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P20436995 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03132210 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P40595424 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 234627_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 234216_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR16004081.hg.1 NR_037184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16004081.hg.1 TC16000291.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000565060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000565060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000565060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000565060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000566535.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000566535.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152628.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152628.hg.1 NR_037184.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152628.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152628.hg.1 NR_037184.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152628.hg.1 TC1600007310.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000499939.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000565060.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000565060.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000565060.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000565060.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000566535.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000566535.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 ENST00000566854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 NR_037184.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007310.hg.1 NR_037184.1 Refseq
          Human Genome U133 Plus 2.0 Array 234216_at - -
          Human Genome U133 Plus 2.0 Array 234627_at - -
          Human Genome U133 Plus 2.0 Array 234216_PM_at - -
          Human Genome U133 Plus 2.0 Array 234627_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234627_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234216_at8 - -
          Human Genome U133 Set 234627_PM_at - -
          Human Genome U133 Set 234216_PM_at - -
          Human Genome U133 Set 234627_at - -
          Human Genome U133 Set 234216_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054417 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054417 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 139798 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline