| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025239.hg.1 |
TC05001843.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025239.hg.1 |
ENST00000450845.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025239.hg.1 |
ENST00000451821.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025239.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025239.hg.1 |
ENST00000509959.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025239.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184686.hg.1 |
TC0500012207.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500184685.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184685.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184685.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184685.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184685.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184685.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184685.hg.1 |
TC0500012207.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000302091.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000450845.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000451821.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000507779.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000509959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000512761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
ENST00000514983.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012207.hg.1 |
NM_194296.1 |
Refseq |