Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00416855_m1
          See other C1ORF194 GE Assays ›
          Gene Symbol:
          C1orf194
          Gene Name:
          chromosome 1 open reading frame 194
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 109105951 - 109113877 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00416855_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C1orf194
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001122961.1 NP_001116433.1 4 - 5 582 111
          XM_005270448.3 XP_005270505.1 3 - 4 485 111
          XM_006710342.3 XP_006710405.1 4 - 5 928 111
          XM_006710343.3 XP_006710406.1 3 - 4 811 111
          XM_011540647.2 XP_011538949.1 4 - 5 959 111
          GenBank mRNA BC127905.1 - 2 - 3 273 111
          BG196327.1 - 2 - 3 245 111
          BG772797.1 - 4 - 5 583 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015186.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002950.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 62698 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 550539 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 822182 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 218394 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 309950 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 387 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P15512 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 5752 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 20350 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 34985 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00144246 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P15512 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P15512 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20479774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00447606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00447606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20479774 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P92752 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P15512 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00144246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001122961_at - -
          Affymetrix Human Gene 1.0 ST Array 7918298 - -
          Affymetrix Human Gene 1.0 ST Array 7918297 - -
          Affymetrix Human Gene 1.0 ST Array 7918299 - -
          Affymetrix Human Gene 1.0 ST Array 7918296 - -
          Affymetrix Human Gene 1.0 ST Array 7918295 - -
          Affymetrix Human Gene 1.1 ST Array 7918294 - -
          Affymetrix HT HG-U133+ PM Array Plate 231044_PM_at - -
          Affymetrix Human Genome U219 Array 11729031_a_at - -
          Affymetrix Human Genome U219 Array 11729031_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01045887.hg.1 TC01002950.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01045887.hg.1 ENST00000369945.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01045887.hg.1 ENST00000369948.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01045887.hg.1 ENST00000369949.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01045887.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100213153.hg.1 TC0100015186.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100213151.hg.1 TC0100015186.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369942.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369945.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369948.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000369949.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000460065.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 ENST00000462402.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 NM_001122961.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015186.hg.1 NM_001122961.1 Refseq
          Human Genome U133 Plus 2.0 Array 231044_at - -
          Human Genome U133 Plus 2.0 Array 231044_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231044_at6 - -
          Human Genome U95 Set 65661_PM_at - -
          Human Genome U95 Set 65661_at - -
          Human Genome U133 Set 231044_PM_at - -
          Human Genome U133 Set 231044_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A049171 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A049171 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 114271 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3251643 NM_001122961.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3251643 BC127905.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3251643 BG196327.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3251643 BG772797.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1652963 NM_001122961.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1652963 BG772797.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1652963 NM_001122961.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1652963 BG772797.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3251643 NM_001122961.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3251643 BC127905.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3251643 BG196327.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3251643 BG772797.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3251643 NM_001122961.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3251643 BC127905.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3251643 BG196327.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3251643 BG772797.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3251643 NM_001122961.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3251643 BC127905.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3251643 BG196327.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3251643 BG772797.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1652963 NM_001122961.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1652963 BG772797.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1652963 NM_001122961.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1652963 BG772797.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1652963 NM_001122961.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1652963 BG772797.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline