| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
1568745_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1568746_PM_a_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052712.hg.1 |
TC01003369.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052712.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052712.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052712.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052714.hg.1 |
TC01003369.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052714.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052714.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052714.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052713.hg.1 |
TC01003369.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052713.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052713.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052716.hg.1 |
TC01003369.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052716.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052716.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052716.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052715.hg.1 |
TC01003369.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052715.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052715.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052715.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222004.hg.1 |
TC0100016059.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222002.hg.1 |
TC0100016059.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222006.hg.1 |
TC0100016059.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016059.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222003.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222003.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222003.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222003.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222003.hg.1 |
TC0100016059.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
ENST00000414848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
ENST00000442358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
NR_033946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222007.hg.1 |
TC0100016059.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1568746_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1568745_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1568746_PM_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1568745_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568746_a_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1568745_at1 |
- |
- |
| Human Genome U95 Set |
47310_PM_at |
- |
- |
| Human Genome U95 Set |
47311_PM_g_at |
- |
- |
| Human Genome U95 Set |
47311_g_at |
- |
- |
| Human Genome U95 Set |
47310_at |
- |
- |
| Human Genome U95 Set |
53329_PM_at |
- |
- |
| Human Genome U95 Set |
53329_at |
- |
- |