| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
7927172 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7927171 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7927170 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
7927169 |
- |
- |
| Affymetrix Human Genome U219 Array |
11748076_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11748076_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003137.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003137.hg.1 |
ENST00000490208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003137.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003137.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003150.hg.1 |
ENST00000431662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003150.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003150.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003150.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000451167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000451167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000490208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000490208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000596267.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000596267.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
ENST00000596267.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003148.hg.1 |
ENST00000431662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003148.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003148.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003148.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003146.hg.1 |
ENST00000431662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003146.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003146.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003146.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003141.hg.1 |
ENST00000431662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003141.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003141.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003141.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003143.hg.1 |
ENST00000431662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003143.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003143.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003143.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003144.hg.1 |
ENST00000431662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003144.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003144.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003144.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003145.hg.1 |
ENST00000431662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003145.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003145.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003145.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003149.hg.1 |
ENST00000437590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003149.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003149.hg.1 |
TC10000275.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
ENST00000451167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
ENST00000490208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
ENST00000451167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
ENST00000490208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150430.hg.1 |
TC1000007421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
ENST00000451167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
ENST00000451167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150436.hg.1 |
TC1000007421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000150439.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150439.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150439.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150439.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150439.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150439.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150439.hg.1 |
TC1000007421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000150442.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150442.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150442.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150442.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150442.hg.1 |
TC1000007421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150440.hg.1 |
TC1000007421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000150441.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150441.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150441.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150441.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150441.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150441.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150441.hg.1 |
TC1000007421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000315429.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000437590.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000451167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000451167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000456416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000490208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000490208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000596267.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000596267.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
ENST00000596267.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007421.hg.1 |
NR_026693.1 |
Refseq |