| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004727.hg.1 |
ENST00000360096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004727.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004727.hg.1 |
TC04000333.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004729.hg.1 |
ENST00000360096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004729.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004729.hg.1 |
TC04000333.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004726.hg.1 |
ENST00000360096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004726.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004726.hg.1 |
TC04000333.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007596.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151976.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151976.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151976.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151976.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151976.hg.1 |
TC0400007596.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400151977.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151977.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151977.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151977.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151977.hg.1 |
TC0400007596.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400151975.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151975.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151975.hg.1 |
TC0400007596.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400151979.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151979.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151979.hg.1 |
ENST00000360096.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151979.hg.1 |
NM_206919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151979.hg.1 |
TC0400007596.hg.1 |
Clariom D TC |