| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001932.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001932.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001932.hg.1 |
TC21000191.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001933.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001933.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001933.hg.1 |
TC21000191.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2100148703.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100148703.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100148703.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100148703.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100148703.hg.1 |
TC2100007221.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100148704.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100148704.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100148704.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100148704.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100148704.hg.1 |
TC2100007221.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100148705.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100148705.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100148705.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100148705.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100148705.hg.1 |
TC2100007221.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
ENST00000413718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007221.hg.1 |
NR_024367.1 |
Refseq |