| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000325.hg.1 |
TC0X000021.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000325.hg.1 |
ENST00000420429.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000325.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000325.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000328.hg.1 |
TC0X000021.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000328.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000328.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006494.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006494.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006494.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006494.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006494.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006494.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000326.hg.1 |
TC0X000021.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000326.hg.1 |
ENST00000420429.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000326.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000326.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144574.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00144574.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144574.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00144574.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144574.hg.1 |
TC0X00006494.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00144575.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00144575.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144575.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00144575.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144575.hg.1 |
TC0X00006494.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00006494.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006494.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006494.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006494.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006494.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006494.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144576.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00144576.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144576.hg.1 |
ENST00000457435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00144576.hg.1 |
NR_038428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144576.hg.1 |
TC0X00006494.hg.1 |
Clariom D TC |
| Human Genome U95 Set |
67762_PM_at |
- |
- |
| Human Genome U95 Set |
67762_at |
- |
- |