Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00417594_g1
          See other C12ORF71 GE Assays ›
          Gene Symbol:
          C12orf71
          Gene Name:
          chromosome 12 open reading frame 71
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 27081057 - 27084127 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00417594_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C12orf71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080406.2 NP_001073875.1 1 - 2 607 91
          NM_001384983.1 NP_001371912.1 2 - 3 672 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010205.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001332.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263831 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16542468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695431 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695464 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695449 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16542465 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16542448 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695448 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695439 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28695496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08263825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16542469 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 455303 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 9850 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3357630 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1413 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1412 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 8645 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P33153394 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P11382527 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3357630 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3357630 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263865 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695480 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695481 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695496 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263851 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695483 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695514 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695470 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695464 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16542469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16542478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695500 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695486 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695492 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695482 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16542465 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263844 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263845 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16542490 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695502 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695466 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695473 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263845 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695470 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16542490 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263865 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16542469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695466 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263851 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695473 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695480 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695483 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695500 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695482 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695486 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695464 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695514 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695492 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695496 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16542465 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16542478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695502 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08263844 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28695481 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P33153394 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263839 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263822 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16542469 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263859 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263840 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263846 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263828 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263855 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263836 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263866 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16542490 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263852 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263844 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263857 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263849 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263862 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16542476 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263860 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263818 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263838 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263833 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263851 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08263816 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11735899_PM_at - -
          Affymetrix Human Genome U219 Array 11735899_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1200010205.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010205.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010205.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010205.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010205.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010205.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010205.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010205.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12017481.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12017481.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12017481.hg.1 TC12001332.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200010217.hg.1 ENST00000538759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010217.hg.1 ENST00000538759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12017477.hg.1 ENST00000398815.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12017477.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12017477.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12017477.hg.1 TC12001332.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12017480.hg.1 ENST00000398815.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12017480.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12017480.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12017480.hg.1 TC12001332.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1200010205.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010205.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010205.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010205.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010205.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010205.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010205.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010205.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177304.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177304.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177304.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177304.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177304.hg.1 TC1200010205.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200177301.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177301.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177301.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177301.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177301.hg.1 TC1200010205.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200177302.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177302.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177302.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177302.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177302.hg.1 TC1200010205.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200010217.hg.1 ENST00000538759.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010217.hg.1 ENST00000538759.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177303.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177303.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177303.hg.1 ENST00000429849.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200177303.hg.1 NM_001080406.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200177303.hg.1 TC1200010205.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047428 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047428 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 126989 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1844083 NM_001080406.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1844083 NM_001080406.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1844083 NM_001080406.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1844083 NM_001080406.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline