Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00417781_m1
          See other LOC100133985 GE Assays ›
          Gene Symbol:
          LOC100133985
          Gene Name:
          uncharacterized LOC100133985
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 70124036 - 70125316 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00417781_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100133985
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024444.1 1 - 2 275 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200012984.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001956.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15253629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15253630 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263201 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15253632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21344192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01263214 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 346727 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3422654 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4569 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4570 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 46243 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3422654 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3422654 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263208 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344225 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263206 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344225 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263208 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263206 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21344232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01263238 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00400235 - -
          Agilent Human 1B Microarray G4111A A_32_P101162 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263224 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263179 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263222 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263230 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263218 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263184 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263216 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263203 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263240 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263208 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263188 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15253629 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263191 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263210 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263199 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15253635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263212 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15253641 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263190 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263232 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263220 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263236 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263214 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15253630 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15253632 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263173 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263182 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263248 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263227 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263183 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263234 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263245 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263195 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01263238 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 230325_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02031098.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02031098.hg.1 NR_024444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02031098.hg.1 TC02001956.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200012984.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012984.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012984.hg.1 NR_024444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012984.hg.1 NR_024444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02031099.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02031099.hg.1 NR_024444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02031099.hg.1 TC02001956.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200194687.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200194687.hg.1 NR_024444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200194687.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200194687.hg.1 NR_024444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200194687.hg.1 TC0200012984.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200194689.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200194689.hg.1 NR_024444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200194689.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200194689.hg.1 NR_024444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200194689.hg.1 TC0200012984.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200012984.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012984.hg.1 ENST00000414141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012984.hg.1 NR_024444.1 Refseq
          Affymetrix Human Clariom D Assay TC0200012984.hg.1 NR_024444.1 Refseq
          Human Genome U133 Plus 2.0 Array 230325_at - -
          Human Genome U133 Plus 2.0 Array 230325_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230325_at4 - -
          Human Genome U95 Set 75086_at - -
          Human Genome U95 Set 75086_PM_at - -
          Human Genome U133 Set 230325_PM_at - -
          Human Genome U133 Set 230325_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013808 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013808 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 143299 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1916599 NR_024444.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1916599 BC015412.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3235101 NR_024444.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3235101 BC015412.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235101 NR_024444.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235101 BC015412.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1916599 NR_024444.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1916599 BC015412.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235101 NR_024444.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235101 BC015412.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235101 NR_024444.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235101 BC015412.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1916599 NR_024444.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1916599 BC015412.2 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 7150274 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_77674 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1916599 NR_024444.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1916599 BC015412.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1916599 NR_024444.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1916599 BC015412.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1916599 NR_024444.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1916599 BC015412.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline