| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008411.hg.1 |
ENST00000416722.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008411.hg.1 |
ENST00000416722.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008411.hg.1 |
NR_103811.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008411.hg.1 |
NR_103811.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008411.hg.1 |
ENST00000416722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008411.hg.1 |
ENST00000416722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008411.hg.1 |
NR_103811.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008411.hg.1 |
NR_103811.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157081.hg.1 |
TC2100008411.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100157081.hg.1 |
ENST00000416722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157081.hg.1 |
NR_103811.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157081.hg.1 |
ENST00000416722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157081.hg.1 |
NR_103811.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157080.hg.1 |
TC2100008411.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100157080.hg.1 |
ENST00000416722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157080.hg.1 |
NR_103811.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157080.hg.1 |
ENST00000416722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157080.hg.1 |
NR_103811.1 |
Refseq |