Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813802 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813831 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223546 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223578 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813797 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813826 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813761 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223543 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813787 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813774 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223535 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813780 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813791 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813823 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813795 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813799 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16454211 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813789 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223592 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223562 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813804 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813796 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813806 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813816 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813785 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813809 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813772 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813800 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223551 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813819 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223591 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813821 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813776 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813814 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813773 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813808 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223571 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813762 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223561 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223587 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223576 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813810 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813781 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223540 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813788 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813825 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813763 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813830 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813832 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813829 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813767 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223532 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813769 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16454218 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223547 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28223580 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07813766 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
847879 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
502047 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
245332 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
349582 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
272240 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
222839 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
133861 |
AB115770.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
133861 |
BC002770.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
133861 |
NM_138383.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
133861 |
XM_001125744.1 |
refseq_mrna_human_XM |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3346716 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_1709 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_1708 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
49114 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P56636977 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P32913406 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P11105807 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P56636986 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3346716 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223540 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223561 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813799 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16454211 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813780 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813774 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813762 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223535 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813767 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813797 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223547 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813779 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813806 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813786 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813795 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813766 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813773 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223562 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813804 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223551 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813785 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813769 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223532 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813776 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813802 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813789 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223546 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813788 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813759 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813772 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813802 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223561 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813776 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813789 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813795 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813786 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223540 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813780 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813797 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813774 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223546 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223535 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223532 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813779 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813759 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813766 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16454211 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813804 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813788 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813772 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813769 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223547 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813799 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813767 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813785 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813773 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223562 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813762 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07813806 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28223551 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19248120 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19248111 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P39366184 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813799 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813774 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813795 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813864 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813778 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813802 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813869 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813776 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813831 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813828 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813872 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813875 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813763 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813769 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813786 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813825 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813810 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813826 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813808 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813759 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813772 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813829 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813861 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813792 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813821 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813758 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813789 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813818 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813780 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16454218 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813817 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813811 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813785 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813819 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813764 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813824 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813868 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813754 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813756 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813766 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813797 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813752 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813806 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813788 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813814 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07813816 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16454211 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16454210 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19248111 |
- |
- |