Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00418278_g1
          See other PET100 GE Assays ›
          Gene Symbol:
          PET100
          Gene Name:
          PET100 homolog
          Gene Aliases:
          C19orf79
          Chromosome Location:
          Chr.19: 7629785 - 7631624 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00418278_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PET100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001171155.1 NP_001164626.1 3 - 4 185 85
          GenBank mRNA AW009489.1 - 1 - 2 85
          BC150496.1 - 3 - 4 163 85
          BU740873.1 - 1 - 2 85
          CD641681.1 - 3 - 4 179 85

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          mitochondrial respiratory chain complex IV assembly

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P391689 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 13493 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55816 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P391689 - -
          Agilent Whole Chr array_18 19 21 A_17_P17104476 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873460 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873473 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873472 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485894 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485889 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873463 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873469 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873465 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873461 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873467 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485895 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873464 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873468 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873471 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873459 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873462 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485897 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873466 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873470 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873474 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31485895 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873467 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31485888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873465 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873471 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17104476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873473 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873462 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873459 - -
          Agilent Whole Chr array_18 19 21 A_17_P17104476 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873465 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485894 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873462 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873467 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873471 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873466 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485895 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873460 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873472 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873468 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485889 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873464 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873474 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873469 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873463 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873461 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873470 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485897 - -
          Agilent Whole Chr array_18 19 21 A_17_P10873473 - -
          Agilent Whole Chr array_18 19 21 A_17_P31485888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31485888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873473 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873465 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31485895 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873467 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873462 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17104476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873471 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10873474 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 226006_PM_at - -
          Human Genome U133 Plus 2.0 Array 226006_at - -
          Human Genome U133 Plus 2.0 Array 226006_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 226006_at11 - -
          Human Genome U95 Set 55586_PM_at - -
          Human Genome U95 Set 55586_at - -
          Human Genome U133 Set 226006_PM_at - -
          Human Genome U133 Set 226006_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A012137 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A003560 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A012137 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A003560 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 181271 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline