| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000559378.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000559378.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000558097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000559378.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000559378.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009839.hg.1 |
ENST00000560662.1 |
Ensembl |