Search
Search

COX8C| Gene Symbol: COX8C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182971.3 | NP_892016.1 | 1 - 2 | 213 | 151 | |
| GenBank mRNA | BC101125.2 | - | 1 - 2 | 179 | 151 | |
| BC101126.2 | - | 1 - 2 | 179 | 151 | ||
Set Membership: |
|