| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170828.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170839.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170839.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170839.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170826.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170837.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170845.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010413.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170829.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170830.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170835.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
ENST00000411439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170831.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170841.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170841.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170841.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
NR_120657.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
NR_120658.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
NR_120659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170846.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000170842.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170842.hg.1 |
NR_120656.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170842.hg.1 |
TC1000010413.hg.1 |
Clariom D TC |