| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005144.hg.1 |
ENST00000502441.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005144.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005144.hg.1 |
TC04000379.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005146.hg.1 |
ENST00000502441.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005146.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005146.hg.1 |
TC04000379.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005148.hg.1 |
ENST00000502441.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005148.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005148.hg.1 |
TC04000379.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005149.hg.1 |
ENST00000502441.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005149.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005149.hg.1 |
TC04000379.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400188380.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188380.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188380.hg.1 |
TC0400012806.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188376.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188376.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188376.hg.1 |
TC0400012806.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188377.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188377.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188377.hg.1 |
TC0400012806.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188378.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188378.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188378.hg.1 |
TC0400012806.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188381.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188381.hg.1 |
NR_033311.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188381.hg.1 |
TC0400012806.hg.1 |
Clariom D TC |