| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000750.hg.1 |
TC0X000055.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000750.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000750.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000749.hg.1 |
TC0X000055.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000749.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000749.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000749.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011273.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
ENST00000456091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177213.hg.1 |
TC0X00011273.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00177214.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177214.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177214.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177214.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177214.hg.1 |
ENST00000419566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177214.hg.1 |
NR_045260.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177214.hg.1 |
TC0X00011273.hg.1 |
Clariom D TC |