| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010031.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171451.hg.1 |
TC0700010031.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010031.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
ENST00000415399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
ENST00000453348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
NR_120508.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
NR_120509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
NR_120510.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700171453.hg.1 |
TC0700010031.hg.1 |
Clariom D TC |