Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00419672_m1
          See other SPINT4 GE Assays ›
          Gene Symbol:
          SPINT4
          Gene Name:
          serine peptidase inhibitor, Kunitz type 4
          Gene Aliases:
          C20orf137, SPINT3, dJ601O1.1
          Chromosome Location:
          Chr.20: 45722349 - 45725696 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00419672_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SPINT4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178455.1 NP_848550.1 2 - 3 310 73
          GenBank mRNA AY372173.1 - 2 - 3 310 73
          AY372174.1 - 2 - 3 310 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007498.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000353.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188293 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188301 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188315 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31836168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11188290 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 913287 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 66862 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 386044 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 208480 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 45125 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 389109 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 250019 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3424302 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3180 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3179 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60529 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03524253 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03524257 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03524256 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3424302 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3424302 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11188290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11188285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11188281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11188281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11188285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11188290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31836160 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03524256 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 30023 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 91312 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03524256 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188293 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188290 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188305 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188281 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188300 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188303 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188297 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188276 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188274 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188285 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188279 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188314 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188283 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188308 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188288 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188316 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11188294 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_178455_at - -
          Affymetrix Human Gene 1.0 ST Array 8063022 - -
          Affymetrix Human Gene 1.0 ST Array 8063020 - -
          Affymetrix Human Gene 1.0 ST Array 8063021 - -
          Affymetrix Human Gene 1.0 ST Array 8063023 - -
          Affymetrix Human Gene 1.1 ST Array 8063019 - -
          Affymetrix Human Genome U219 Array 11734351_PM_at - -
          Affymetrix Human Genome U219 Array 11734351_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR20005070.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20005070.hg.1 NM_178455.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20005070.hg.1 TC20000353.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000007498.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007498.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007498.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007498.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007498.hg.1 NM_178455.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007498.hg.1 NM_178455.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007498.hg.1 NM_178455.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000152216.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2000152216.hg.1 NM_178455.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000152216.hg.1 TC2000007498.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000152216.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2000152216.hg.1 NM_178455.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007498.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000007498.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000007498.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000007498.hg.1 ENST00000279058.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000007498.hg.1 NM_178455.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007498.hg.1 NM_178455.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007498.hg.1 NM_178455.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018380 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018380 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1723279 NM_178455.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1723279 AY372173.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1723279 AY372174.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723279 AY372173.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723279 AY372174.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1723279 NM_178455.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1723279 AY372173.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1723279 AY372174.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1723279 AY372173.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1723279 AY372174.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1723279 AY372173.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1723279 AY372174.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1723279 AY372173.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1723279 AY372174.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1723279 AY372173.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1723279 AY372174.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1723279 AY372173.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1723279 AY372174.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1723279 AY372173.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1723279 AY372174.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1723279 AY372173.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1723279 AY372174.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1723279 NM_178455.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1723279 AY372173.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1723279 AY372174.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline