Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00419965_g1
          See other KU-MEL-3 GE Assays ›
          Gene Symbol:
          KU-MEL-3 NEFM
          Gene Name:
          uncharacterized LOC497048
          neurofilament medium chain
          Gene Aliases:
          -
          NEF3, NF-M, NFM
          Chromosome Location:
          Chr.6: 4610565 - 4611919 on Build GRCh38
          Chromosome Location:
          Chr.8: 24913761 - 24919093 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00419965_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KU-MEL-3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_122107.1 1 - 2 246 69
          GenBank mRNA AY870321.1 - 1 - 2 199 69
          BC130651.1 - 1 - 2 94 69
          BC130655.1 - 1 - 2 99 69
          Gene Symbol: NEFM
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF181990.1 - 1 - 2 199 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006779.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708802 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04522006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04522005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04522015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708810 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15779720 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04522012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708849 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521931 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04522003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04521937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708803 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24708838 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14148 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3267195 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 46935 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 97884 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8116709 - -
          Affymetrix Human Gene 1.0 ST Array 8116708 - -
          Affymetrix Human Gene 1.1 ST Array 8116707 - -
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146583.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146583.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146583.hg.1 TC0600006779.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146579.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146579.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146579.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146579.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146579.hg.1 TC0600006779.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146581.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146581.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146581.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146581.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146581.hg.1 TC0600006779.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146582.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146582.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146582.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146582.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146582.hg.1 TC0600006779.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 ENST00000380106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006779.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146584.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146584.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146584.hg.1 TC0600006779.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146578.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146578.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146578.hg.1 TC0600006779.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146577.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146577.hg.1 NR_122107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600146577.hg.1 TC0600006779.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 243989_at - -
          Human Genome U133 Plus 2.0 Array 243989_PM_at - -
          Human Genome U95 Set 59529_PM_at - -
          Human Genome U95 Set 59529_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 186127 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1704916 AY870321.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1704916 AY870321.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1704916 AY870321.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1704916 AY870321.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1704916 AY870321.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1704916 AY870321.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0