Search
Search

LOC729296| Gene Symbol: LOC729296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109916.1 | 2 - 3 | 438 | 70 | ||
| NR_109917.1 | 2 - 3 | 438 | 70 | |||
| NR_109918.1 | 2 - 3 | 438 | 70 | |||
| GenBank mRNA | AW082248.1 | - | 3 - 4 | 70 | ||
| DB026168.1 | - | 2 - 3 | 416 | 70 | ||
| DB051395.1 | - | 2 - 3 | 438 | 70 | ||
| HY046567.1 | - | 2 - 3 | 252 | 70 | ||
Set Membership: |
|